missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpiperazine, 97%, Thermo Scientific™

Catalog Number p-3758674
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
92-54-6
C10H14N2
162.24
MFCD00005957
YZTJYBJCZXZGCT-UHFFFAOYSA-N
phenylpiperazine, n-phenylpiperazine, piperazine, 1-phenyl, 1-phenyl-piperazine, n-phenyldiethylenediamine, 1-fenylpiperazin czech, unii-j9225cbi7d, n-phenyl piperazine, 1-phenyl piperazine, ccris 4334
7096
1-phenylpiperazine
C1CN(CCN1)C2=CC=CC=C2
This item is not returnable. View return policy
92-54-6
C10H14N2
162.24
MFCD00005957
YZTJYBJCZXZGCT-UHFFFAOYSA-N
phenylpiperazine, n-phenylpiperazine, piperazine, 1-phenyl, 1-phenyl-piperazine, n-phenyldiethylenediamine, 1-fenylpiperazin czech, unii-j9225cbi7d, n-phenyl piperazine, 1-phenyl piperazine, ccris 4334
7096
1-phenylpiperazine
C1CN(CCN1)C2=CC=CC=C2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 18.8°C
Color Colorless to Yellow
Density 1.0620g/mL
Boiling Point 286.0°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. (Aniline) (GC)
Packaging Glass bottle
Refractive Index 1.5865 to 1.5885
Quantity 25 g
UN Number 2927
Specific Gravity 1.062
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 162.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Phenylpiperazine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'