missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclopentanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-181587
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
77-55-4
C12H14O2
190.24
MFCD00001372
RHPCYZLXNNRRMB-UHFFFAOYSA-N
1-phenylcyclopentanecarboxylic acid, 1-phenyl-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopentanecarboxylic acid, 1-phenylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-phenyl, pubchem22923, acmc-209pao, phenylcyclopentanecarboxylic acid, phenylcyclopentane carboxylic acid
66167
1-phenylcyclopentane-1-carboxylic acid
C1CCC(C1)(C2=CC=CC=C2)C(=O)O
This item is not returnable. View return policy
77-55-4
C12H14O2
190.24
MFCD00001372
RHPCYZLXNNRRMB-UHFFFAOYSA-N
1-phenylcyclopentanecarboxylic acid, 1-phenyl-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopentanecarboxylic acid, 1-phenylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-phenyl, pubchem22923, acmc-209pao, phenylcyclopentanecarboxylic acid, phenylcyclopentane carboxylic acid
66167
1-phenylcyclopentane-1-carboxylic acid
C1CCC(C1)(C2=CC=CC=C2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 157°C to 161°C
Color Brown-Gray to Cream
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5C5H8CO2H
Quantity 5 g
Beilstein 09, II, 418
Formula Weight 190.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Phenylcyclopentanecarboxylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenylcyclopentanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'