missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Phenyl-3-n-propyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64855.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15472249

  • £119.78 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

1-Phenyl-3-n-propyl-2-thiohydantoin
234.32
MFCD26967927
1-Phenyl-3-n-propyl-2-thioxo-4-imidazolidinone

Specifications

1-Phenyl-3-n-propyl-2-thiohydantoin
Solid
234.32
1-Phenyl-3-n-propyl-2-thioxo-4-imidazolidinone
1g
95%
MFCD26967927
Videos
SDS