missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar 1-Phenyl-3-[3-(trifluoromethyl)phenyl]urea, 95%

Brand:  Alfa Aesar H61601.03

1-Phenyl-3-[3-(trifluoromethyl)phenyl]urea, 95%

Code : NEW

Additional Details : CAS Number : 4300-46-3 Weight : 0.00100kg

Product Code. 15478158

  • £73.56 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket