missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-thiourea, 97%, ACROS Organics™

£13.40 - £106.60

Chemical Identifiers

CAS 103-85-5
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.21
MDL Number MFCD00004933
InChI Key FULZLIGZKMKICU-UHFFFAOYSA-N
Synonym n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
PubChem CID 676454
ChEBI CHEBI:46261
IUPAC Name phenylthiourea
SMILES C1=CC=C(C=C1)NC(=S)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10616792
SDS View Documents
Acros Organics
207250050
5g Glass bottle £13.40
5g View alternative products
Save Up to 
»
In stock, ready to ship: 3 Please to purchase this item. Need a web account? Register with us today!
10107703
SDS View Documents
Acros Organics
207250250
25g Glass bottle £41.98
25g View alternative products
Save Up to 
»
In stock, ready to ship: 2 Please to purchase this item. Need a web account? Register with us today!
10036383
SDS View Documents
Acros Organics
207251000
100g Glass bottle £106.60
100g View alternative products
Save Up to 
»
In stock, ready to ship: 3 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

103-85-5
152.21
FULZLIGZKMKICU-UHFFFAOYSA-N
676454
phenylthiourea
C7H8N2S
MFCD00004933
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
CHEBI:46261
C1=CC=C(C=C1)NC(=S)N

Specifications

1-Phenyl-2-thiourea
Glass bottle
148°C to 150°C
103-85-5
100.0
C7H8N2S
MFCD00004933
15, 7424
Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol,aq.ether,naoh,ethanol
C1=CC=C(C=C1)NC(=S)N
152.21
CHEBI:46261
Crystalline Powder
Authentic
White to Beige
5g
96.0
96% min. (HPLC)
C6H5NHCSNH2
12, 388
n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
FULZLIGZKMKICU-UHFFFAOYSA-N
phenylthiourea
676454
152.21
97%