missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-propanol, 99%, ACROS Organics™

Brand:  Acros Organics 183060250

Code : 29

Additional Details : CAS Number : 14898-87-4 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10661784

  • £55.40 / 25g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

1-Phenyl-2-propanol
98.5
≥98.5%
C9H12O
136.19
α-Methylphenethyl alcohol
14898-87-4
100.0
99%
C6H5CH2CH(OH)CH3
MFCD00004540

Specifications

1-Phenyl-2-propanol
Clear colorless
14898-87-4
98.5
Glass bottle
Liquid
99%
C6H5CH2CH(OH)CH3
MFCD00004540
0.9900g/mL
25g
Authentic
100.0
1.5200 to 1.5220 (20°C, 589nm)
≥98.5%
C9H12O
136.19
α-Methylphenethyl alcohol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Phenyl-2-propanol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS