missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1H-pyrazole-5-carbaldehyde, 95+%, Thermo Scientific™

Catalog Number p-4510275
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
132274-70-5
C10H8N2O
172.187
OQORFMABOZEDBL-UHFFFAOYSA-N
1-phenyl-1h-pyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1-phenyl, 1-phenylpyrazole-5-carbaldehyde, 1-phenyl-1h-pyrazole-5-carboxaldehyde
2776567
2-phenylpyrazole-3-carbaldehyde
C1=CC=C(C=C1)N2C(=CC=N2)C=O
This item is not returnable. View return policy
132274-70-5
C10H8N2O
172.187
OQORFMABOZEDBL-UHFFFAOYSA-N
1-phenyl-1h-pyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1-phenyl, 1-phenylpyrazole-5-carbaldehyde, 1-phenyl-1h-pyrazole-5-carboxaldehyde
2776567
2-phenylpyrazole-3-carbaldehyde
C1=CC=C(C=C1)N2C(=CC=N2)C=O
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 172.19
Percent Purity 95%
Chemical Name or Material 1-Phenyl-1H-pyrazole-5-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'