missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-trimethylsiloxyethylene, 95%, Thermo Scientific Chemicals

Catalog Number p-7044215
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13735-81-4
C11H16OSi
192.333
MFCD00008582
AFFPCIMDERUIST-UHFFFAOYSA-N
1-phenyl-1-trimethylsiloxyethylene, trimethyl 1-phenylvinyl oxy silane, 1-phenyl-1-trimethylsilyloxyethylene, 1-phenyl-1-trimethylsiloxy ethylene, 1-phenyl-1-trimethylsilyloxy ethylene, benzene, 1-trimethylsilyl oxy ethenyl, silane, trimethyl 1-phenylethenyl oxy, acmc-209ccf
117406
trimethyl(1-phenylethenoxy)silane
C[Si](C)(C)OC(=C)C1=CC=CC=C1
This item is not returnable. View return policy
13735-81-4
C11H16OSi
192.333
MFCD00008582
AFFPCIMDERUIST-UHFFFAOYSA-N
1-phenyl-1-trimethylsiloxyethylene, trimethyl 1-phenylvinyl oxy silane, 1-phenyl-1-trimethylsilyloxyethylene, 1-phenyl-1-trimethylsiloxy ethylene, 1-phenyl-1-trimethylsilyloxy ethylene, benzene, 1-trimethylsilyl oxy ethenyl, silane, trimethyl 1-phenylethenyl oxy, acmc-209ccf
117406
trimethyl(1-phenylethenoxy)silane
C[Si](C)(C)OC(=C)C1=CC=CC=C1

1-Phenyl-1-trimethylsiloxyethylene has been used in the synthesis of β-amino ketones in water via Mannich-type reaction and cis-2,6-disubstituted dihydropyrans via three-component, one-pot cascade reaction. It is also used as chemical additives and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1-trimethylsiloxyethylene has been used in the synthesis of β-amino ketones in water via Mannich-type reaction and cis-2,6-disubstituted dihydropyrans via three-component, one-pot cascade reaction. It is also used as chemical additives and intermediates.

Solubility
Reacts with water.

Notes
Store at 4°C. Moisture Sensitive. Store under inert gas. Incompatible materials are Oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 53°C to 54°C (1mmHg)
Quantity 1 g
Beilstein 1306914
Flash Point 78°C (172°F)
Solubility Information Reacts with water.
Formula Weight 192.34
Percent Purity 97%
Refractive Index 1.502
Sensitivity Moisture sensitive
Density 0.938
Chemical Name or Material 1-Phenyl-1-trimethylsiloxyethylene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'