missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclopropanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-181560
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6120-95-2
C10H10O2
162.19
MFCD00001288
IWWCCNVRNHTGLV-UHFFFAOYSA-N
1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid
80206
1-phenylcyclopropane-1-carboxylic acid
C1CC1(C2=CC=CC=C2)C(=O)O
This item is not returnable. View return policy
6120-95-2
C10H10O2
162.19
MFCD00001288
IWWCCNVRNHTGLV-UHFFFAOYSA-N
1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid
80206
1-phenylcyclopropane-1-carboxylic acid
C1CC1(C2=CC=CC=C2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 84°C to 87°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C3H4CO2H
Quantity 10 g
Formula Weight 162.19
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1-Phenyl-1-cyclopropanecarboxylic acid, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'