missing translation for 'onlineSavingsMsg'
Learn More

1-Pentene, 97%, Thermo Scientific Chemicals

Catalog Number 10464463
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
109-67-1
C5H10
70.14
MFCD00003567
YWAKXRMUMFPDSH-UHFFFAOYSA-N
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
8004
pent-1-ene
CCCC=C
109-67-1
C5H10
70.14
MFCD00003567
YWAKXRMUMFPDSH-UHFFFAOYSA-N
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
8004
pent-1-ene
CCCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -165.0°C
Color Colorless
Density 0.6400g/mL
Boiling Point 30.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3CH2CH2CH=CH2
Refractive Index 1.3704 to 1.3724
Quantity 500 mL
Beilstein 01,21
Merck Index 15,7234
Specific Gravity 0.64
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether and beznene, soluble in most organic solvents
Viscosity 0.202 mPa.s (20°C)
Formula Weight 70.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Pentene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'