Learn More
1-Pentanol, ACS, 99+%, Thermo Scientific Chemicals
Description
1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -79°C |
Color | Colorless |
Density | 0.812 g/mL |
Boiling Point | 136°C to 138°C |
Flash Point | 49°C (120°F) |
Odor | Mild |
Linear Formula | CH3(CH2)4OH |
Refractive Index | 1.4103 |
Quantity | 4 L |
UN Number | UN1105 |
Show More |
RUO – Research Use Only