missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals

Catalog Number 11914571
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
22767-49-3
C5H11NaO3S
174.19
MFCD00007541,MFCD00149548
ROBLTDOHDSGGDT-UHFFFAOYSA-M
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
23664617
sodium;pentane-1-sulfonate
[Na+].CCCCCS([O-])(=O)=O
22767-49-3
C5H11NaO3S
174.19
MFCD00007541,MFCD00149548
ROBLTDOHDSGGDT-UHFFFAOYSA-M
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
23664617
sodium;pentane-1-sulfonate
[Na+].CCCCCS([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY
Absorbance 0.02 max. (C=0.5 M in water) at 260nm, 0.04 max. (C=0.5 M in water) at 230nm, 0.06 max. C=0.5 M in water at 220nm, 0.1 max. (C=0.5 M in water) at 210nm
Melting Point >300.0°C
Color White
Loss on Drying 2% (150°C) max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula CH3(CH2)4SO3Na
Quantity 500 g
Solubility Information Solubility in water: soluble.
Formula Weight 174.2
Percent Purity 99+%
Grade Ion Pair Chromatography
Physical Form Crystalline Powder
Chemical Name or Material 1-Pentanesulfonic acid, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • 1-Pentanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'