missing translation for 'onlineSavingsMsg'
Learn More

1-Octyne, 99%, Thermo Scientific Chemicals

Catalog Number 10542431
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
629-05-0
C8H14
110.20
MFCD00009546
UMIPWJGWASORKV-UHFFFAOYSA-N
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
12370
oct-1-yne
CCCCCCC#C
This item is not returnable. View return policy
629-05-0
C8H14
110.20
MFCD00009546
UMIPWJGWASORKV-UHFFFAOYSA-N
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
12370
oct-1-yne
CCCCCCC#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -80°C
Color Colorless to Yellow
Density 0.7400g/mL
Boiling Point 127°C to 128°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)5C∽CH
Refractive Index 1.4150 to 1.417
Quantity 25 mL
Beilstein 01, 258
Specific Gravity 0.74
Solubility Information Solubility in water: 0.024g/L (25°C)
Formula Weight 110.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Octyne
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Octyne
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'