missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Octylphosphonic acid, 98%

£31.30 - £124.00

Chemical Identifiers

CAS 4724-48-5
Molecular Formula C8H19O3P
Molecular Weight (g/mol) 194.211
MDL Number MFCD00015841
InChI Key NJGCRMAPOWGWMW-UHFFFAOYSA-N
Synonym n-octylphosphonic acid, 1-octylphosphonic acid, phosphonic acid, octyl, phosphonic acid, p-octyl, unii-95c2aw87bo, octylphosphonicacid, acmc-209uni, p-octyl-phosphonic acid, octylphosphonic acid, dsstox_cid_24679
PubChem CID 78452
IUPAC Name octylphosphonic acid
SMILES CCCCCCCCP(=O)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307967 SDS View Documents Alfa Aesar™
020651.03
1g £31.30
1g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
11397957 SDS View Documents Alfa Aesar™
020651.06
5g £124.00
5g View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

4724-48-5
194.211
NJGCRMAPOWGWMW-UHFFFAOYSA-N
78452
CCCCCCCCP(=O)(O)O
C8H19O3P
MFCD00015841
n-octylphosphonic acid, 1-octylphosphonic acid, phosphonic acid, octyl, phosphonic acid, p-octyl, unii-95c2aw87bo, octylphosphonicacid, acmc-209uni, p-octyl-phosphonic acid, octylphosphonic acid, dsstox_cid_24679
octylphosphonic acid

Specifications

1-Octylphosphonic acid
1g
C8H19O3P
1764242
Slightly soluble in organic solvents and water.
CCCCCCCCP(=O)(O)O
194.211
194.21
98%
100°C to 102°C
4724-48-5
MFCD00015841
n-octylphosphonic acid, 1-octylphosphonic acid, phosphonic acid, octyl, phosphonic acid, p-octyl, unii-95c2aw87bo, octylphosphonicacid, acmc-209uni, p-octyl-phosphonic acid, octylphosphonic acid, dsstox_cid_24679
NJGCRMAPOWGWMW-UHFFFAOYSA-N
octylphosphonic acid
78452
Crystalline