Learn More
1-Octylphosphonic acid, 98%, Thermo Scientific Chemicals
Description
1-Octylphosphonic acid is a basic building block and used in the preparation of various pharmaceutical compounds and chemical products. It is supported with calcium hydroxyapatite and used in the dental field as corrosion inhibitor and organic building block. It acts as a corrosion inhibitor in metallic household surfaces such as faucets and drains as well as prevents the rusting of can or dispenser. It is also used in the synthesis of octyl-phosphonic acid dimethyl ester by reacting with iodomethane in presence of CS2 CO3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 100°C to 102°C |
Quantity | 5 g |
Beilstein | 1764242 |
Solubility Information | Slightly soluble in organic solvents and water. |
Formula Weight | 194.21 |
Percent Purity | 98% |
Physical Form | Crystalline |
Chemical Name or Material | 1-Octylphosphonic acid |
RUO – Research Use Only