missing translation for 'onlineSavingsMsg'
Learn More

1-Octyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7021954
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4430-45-9
C9H17NS
171.30
MFCD00041139
YEZHGQZHWKJPCM-UHFFFAOYSA-N
octyl isothiocyanate, n-octyl isothiocyanate, octylisothiocyanate, 1-octyl isothiocyanate, octanisothiocyanate, 1-isothiocyanatooctan, octane, 1-isothiocyanato, isothiocyanic acid, octyl ester, n-octylisothiocyanate, acmc-1ajcy
78161
1-isothiocyanatooctane
CCCCCCCCN=C=S
This item is not returnable. View return policy
4430-45-9
C9H17NS
171.30
MFCD00041139
YEZHGQZHWKJPCM-UHFFFAOYSA-N
octyl isothiocyanate, n-octyl isothiocyanate, octylisothiocyanate, 1-octyl isothiocyanate, octanisothiocyanate, 1-isothiocyanatooctan, octane, 1-isothiocyanato, isothiocyanic acid, octyl ester, n-octylisothiocyanate, acmc-1ajcy
78161
1-isothiocyanatooctane
CCCCCCCCN=C=S

1-Octyl isothiocyanate can be used to produce 1-(6-methyl-quinolin-2-yl)-3-octyl-thiourea. It is used as intermediates in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octyl isothiocyanate can be used to produce 1-(6-methyl-quinolin-2-yl)-3-octyl-thiourea. It is used as intermediates in pharmaceutical industry.

Solubility
It hydrolyzes in water.

Notes
It is moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
TRUSTED_SUSTAINABILITY
Density 0.88
Boiling Point 124°C to 126°C (7 mmHg)
Flash Point 69°C (156°F)
Odor Pungent
Refractive Index 1.49
Quantity 5 g
UN Number UN2922
Beilstein 1760783
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 171.31
Percent Purity 97%
Chemical Name or Material 1-Octyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'