missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-ol, 98%, Thermo Scientific Chemicals

Catalog Number p-7081898
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
3391-86-4
C8H16O
128.215
MFCD00004589
VSMOENVRRABVKN-UHFFFAOYSA-N
1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
18827
CHEBI:34118
oct-1-en-3-ol
CCCCCC(C=C)O
This item is not returnable. View return policy
3391-86-4
C8H16O
128.215
MFCD00004589
VSMOENVRRABVKN-UHFFFAOYSA-N
1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
18827
CHEBI:34118
oct-1-en-3-ol
CCCCCC(C=C)O

Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point -49°C
Density 0.833
Boiling Point 173°C to 175°C
Flash Point 61°C (141°F)
Odor Characteristic
Refractive Index 1.437
Quantity 10 g
Beilstein 1744110
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 1-Octen-3-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'