missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, natural, 98%, Thermo Scientific Chemicals

Catalog Number p-7052539
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
This item is not returnable. View return policy
111-87-5
C8H18O
130.23
MFCD00002988
KBPLFHHGFOOTCA-UHFFFAOYSA-N
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
957
CHEBI:16188
octan-1-ol
CCCCCCCCO
This item is not returnable. View return policy
111-87-5
C8H18O
130.23
MFCD00002988
KBPLFHHGFOOTCA-UHFFFAOYSA-N
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
957
CHEBI:16188
octan-1-ol
CCCCCCCCO

1-Octanol is a synthetic reagent used to produce esters. It inhibits T-type calcium channels and is used to determine the partition coefficient between compounds and the cytosol. It is used to evaluate the lipophilicity of pharmaceutical products. It is also used as a solvent for protective coatings, waxes, and oils, and as a raw material for plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanol is a synthetic reagent used to produce esters. It inhibits T-type calcium channels and is used to determine the partition coefficient between compounds and the cytosol. It is used to evaluate the lipophilicity of pharmaceutical products. It is also used as a solvent for protective coatings, waxes, and oils, and as a raw material for plasticizers.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY
Melting Point -16°C
Color Colorless
Density 0.825 g/mL
Boiling Point 193°C to 195°C
Flash Point 81°C (178°F)
Odor Sweet
Refractive Index 1.4295
Quantity 500 mL
Solubility Information Not miscible in water.
Formula Weight 130.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Octanol, Natural
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'