missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic acid, sodium salt monohydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-4509448
29.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
207596-29-0
C8H19NaO4S
234.29
MFCD00149551
MBURIAHQXJQKRE-UHFFFAOYSA-M
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate
23666339
sodium;octane-1-sulfonate;hydrate
O.[Na+].CCCCCCCCS([O-])(=O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
207596-29-0
C8H19NaO4S
234.29
MFCD00149551
MBURIAHQXJQKRE-UHFFFAOYSA-M
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate
23666339
sodium;octane-1-sulfonate;hydrate
O.[Na+].CCCCCCCCS([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)7SO3Na·H2O
Quantity 5 g
Beilstein 04,III,25
Solubility Information Solubility in water: soluble. Other solubilities: 4g/l methanol
Formula Weight 234.3
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material 1-Octanesulfonic acid, sodium salt monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'