missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic Acid Sodium Salt, HPLC Grade, J.T. Baker™

Catalog Number p-5383265
366.70 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
25 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
5324-84-5
C8H17NaO3S
216.27
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
23669624
sodium;octane-1-sulfonate
[Na+].CCCCCCCCS([O-])(=O)=O
This item is not returnable. View return policy
5324-84-5
C8H17NaO3S
216.27
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
23669624
sodium;octane-1-sulfonate
[Na+].CCCCCCCCS([O-])(=O)=O

High purity solvents for use in analytical and preparative separations

    For Ion-Pair Chromatography of Basic Compounds
Packaging Wide Mouth Amber Glass Bottle
MDL Number MFCD00007544
Quantity 25 g
Specific Gravity 1L = 0.00kg
Formula Weight 216.27
Grade HPLC
Chemical Name or Material 1-Octanesulfonic Acid Sodium Salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'