missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octadecanol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
500 g
2500 g
Description
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.
Solubility
Insoluble in water. Soluble in alcohol, acetone, ether, benzene, chloroform.
Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.
Solubility
Insoluble in water. Soluble in alcohol, acetone, ether, benzene, chloroform.
Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
Specifications
Melting Point | 56°C to 60°C |
Boiling Point | 210°C (15 mmHg) |
Flash Point | 174°C (345°F) |
Assay Percent Range | 97% |
Quantity | 250 g |
Beilstein | 1362907 |
Merck Index | 14,8805 |
Solubility Information | Insoluble in water. Soluble in alcohol,acetone,ether,benzene,chloroform. |
Formula Weight | 270.5 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'