missing translation for 'onlineSavingsMsg'
Learn More

1-Nonyne 98%, ACROS Organics™

Brand:  Acros Organics 392750050

Code : 29

Additional Details : CAS Number : 9/3/3452 Weight : 0.06000kg Transport : UN number : 3295 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10075143

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

1-Nonyne
100.0
98%
CH3(CH2)6CCH
MFCD00009567
97.5
≥97.5%
C9H16
124.23

Specifications

1-Nonyne
34°C
Clear colorless to pale yellow
5g
100.0
Glass bottle
Liquid
98%
CH3(CH2)6CCH
MFCD00009567
0.7570g/mL
150.0° to 151.0°C
−50.0°C
97.5
Authentic
1.4210 to 1.4230 (20°C, 589nm)
≥97.5%
C9H16
124.23
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Nonyne

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS