missing translation for 'onlineSavingsMsg'
Learn More

1-Nonanol, 99%, Thermo Scientific Chemicals

Catalog Number p-7020525
13.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
143-08-8
C9H20O
144.258
MFCD00002990
ZWRUINPWMLAQRD-UHFFFAOYSA-N
1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
8914
CHEBI:35986
nonan-1-ol
CCCCCCCCCO
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
143-08-8
C9H20O
144.258
MFCD00002990
ZWRUINPWMLAQRD-UHFFFAOYSA-N
1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
8914
CHEBI:35986
nonan-1-ol
CCCCCCCCCO

The primary use of nonanol is in the manufacture of artificial lemon oil. 1-Nonanol is mainly used in man-made rose essence, plasticizer, surfactant. 1-Nonanol was used as a reference material in the determination of airborne spore levels. 1-Nonanol inhibited the bacterial luciferase (BL) reaction in a dose-dependent manner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The primary use of nonanol is in the manufacture of artificial lemon oil. 1-Nonanol is mainly used in man-made rose essence, plasticizer, surfactant. 1-Nonanol was used as a reference material in the determination of airborne spore levels. 1-Nonanol inhibited the bacterial luciferase (BL) reaction in a dose-dependent manner.

Solubility
Soluble in water (1 g/L) at 20°C, ether, ethanol, acetone, chloroform, and mineral oil.

Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY
Melting Point -7°C
Density 0.827
Boiling Point 214°C to 216°C
Flash Point 75°C (167°F)
Odor Pleasant
Linear Formula CH3(CH2)8OH
Refractive Index 1.433
Quantity 100 g
UN Number UN3082
Beilstein 969213
Merck Index 14,6679
Solubility Information Soluble in water (1g/L) at 20°C,ether,ethanol,acetone,chloroform,and mineral oil.
Formula Weight 144.26
Percent Purity 99%
Chemical Name or Material 1-Nonanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'