missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt, 90+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt is the most sensitive determination of trace cobalt reagents used for determination of cobalt and potassium-sensitive reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt is the most sensitive determination of trace cobalt reagents used for determination of cobalt and potassium-sensitive reagents.
Solubility
Slightly soluble in methanol and ethanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt is the most sensitive determination of trace cobalt reagents used for determination of cobalt and potassium-sensitive reagents.
Solubility
Slightly soluble in methanol and ethanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
Melting Point | >300°C |
Quantity | 25 g |
Beilstein | 3643930 |
Merck Index | 14,6644 |
Solubility Information | Slightly soluble in methanol and ethanol. |
IUPAC Name | disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate |
Formula Weight | 377.26 |
Percent Purity | ≥90% |
Chemical Name or Material | 1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'