missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Nitro-2-(trifluoromethoxy)benzene, 97%

Brand:  Alfa Aesar™ H61672.22

Code : NEW

Additional Details : CAS Number : 1644-88-8 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15428238

  • £215.18 / 100g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

1-Nitro-2-(trifluoromethoxy)benzene
207.11
2-(Trifluoromethoxy)nitrobenzene
1644-88-8
MFCD00041010

Specifications

1-Nitro-2-(trifluoromethoxy)benzene
1644-88-8
97%
MFCD00041010
100g
Solid
207.11
2-(Trifluoromethoxy)nitrobenzene
Videos
SDS