Learn More
1-Naphthylacetic acid, 95%, may cont. up to 5% 2-isomer, Thermo Scientific Chemicals
Description
1-Naphthylacetic acid is used as a plant growth regulator and for plant tissue culture. It is used for the vegetative propagation of plants from stem and leaf cutting, abscission promoter and substrate for auxin inducers of specific plant enzymes. Further, it stimulates root formation in cuttings of woody and herbaceous plants. It is also used to prevent premature flower and fruit drop. It is also employed to control regrowth of tree sprouts after trimminguse. In addition to this, it is used in ornaments and serves as an ingredient in many commercial plant rooting horticultural products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Naphthylacetic acid is used as a plant growth regulator and for plant tissue culture. It is used for the vegetative propagation of plants from stem and leaf cutting, abscission promoter and substrate for auxin inducers of specific plant enzymes. Further, it stimulates root formation in cuttings of woody and herbaceous plants. It is also used to prevent premature flower and fruit drop. It is also employed to control regrowth of tree sprouts after trimminguse. In addition to this, it is used in ornaments and serves as an ingredient in many commercial plant rooting horticultural products.
Solubility
Slightly soluble in water, ethanol, acetone, chloroform and ether.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | 126°C to 133°C |
Quantity | 50 g |
Beilstein | 1308415 |
Merck Index | 14,6371 |
Solubility Information | Slightly soluble in water,ethanol,acetone,chloroform and ether. |
Formula Weight | 186.21 |
Percent Purity | 96% |
Concentration or Composition (by Analyte or Components) | May contain up to 4% 2-isomer |
Chemical Name or Material | 1-Naphthylacetic acid |
RUO – Research Use Only