missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl phosphate, calcium salt trihydrate, 97%, Thermo Scientific Chemicals

Catalog Number 10255911
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 17805-69-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 192°C to 193°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Complexometry)
Packaging Glass bottle
Molecular Formula C20H16CaO8P2·3H2O
MDL Number MFCD00149488
Quantity 5 g
Synonym calcium naphthalen-1-yl phosphate trihydrate, calcium trihydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate calcium salt trihydrate, c10h7po4.ca.3h2o, 1-naphthyl phosphate calcium trihydrate
InChI Key XGDPLNLBXXLFDL-UHFFFAOYSA-L
SMILES C1=CC=C2C(=C1)C=CC=C2OP(=O)([O-])[O-].O.O.O.[Ca+2]
IUPAC Name calcium;naphthalen-1-yl phosphate;trihydrate
Molecular Weight (g/mol) 540.41
PubChem CID 51346138
Formula Weight 540.41
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Naphthyl phosphate, calcium salt trihydrate, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'