missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-170984
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
551-06-4
C11H7NS
185.25
MFCD00003882
JBDOSUUXMYMWQH-UHFFFAOYSA-N
1-naphthyl isothiocyanate, 1-naphthylisothiocyanate, anit, kesscocide, alpha-naphthyl isothiocyanate, naphthalene, 1-isothiocyanato, naphthalene, isothiocyanato, isothiocyanic acid, 1-naphthyl ester, 1-naftylisothiokyanat, isothiocyanic acid 1-naphthyl ester
11080
CHEBI:35455
1-isothiocyanatonaphthalene
C1=CC=C2C(=C1)C=CC=C2N=C=S
This item is not returnable. View return policy
551-06-4
C11H7NS
185.25
MFCD00003882
JBDOSUUXMYMWQH-UHFFFAOYSA-N
1-naphthyl isothiocyanate, 1-naphthylisothiocyanate, anit, kesscocide, alpha-naphthyl isothiocyanate, naphthalene, 1-isothiocyanato, naphthalene, isothiocyanato, isothiocyanic acid, 1-naphthyl ester, 1-naftylisothiokyanat, isothiocyanic acid 1-naphthyl ester
11080
CHEBI:35455
1-isothiocyanatonaphthalene
C1=CC=C2C(=C1)C=CC=C2N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 55.5°C to 57.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C10H7NCS
Quantity 5 g
Beilstein 12, 1244
Fieser 01,718
Merck Index 15, 6493
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ether, benzene, hot alcohol,, acetone and ccl4
Formula Weight 185.25
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Naphthyl isothiocyanate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Naphthyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'