missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl butyrate, 98%, Thermo Scientific Chemicals

Catalog Number p-170965
13.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3121-70-8
C14H14O2
214.26
MFCD00003923
XIVJNRXPRQKFRZ-UHFFFAOYSA-N
1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik
76571
naphthalen-1-yl butanoate
CCCC(=O)OC1=CC=CC2=CC=CC=C21
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3121-70-8
C14H14O2
214.26
MFCD00003923
XIVJNRXPRQKFRZ-UHFFFAOYSA-N
1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik
76571
naphthalen-1-yl butanoate
CCCC(=O)OC1=CC=CC2=CC=CC=C21

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.0900g/mL
Boiling Point 125°C to 130°C (1.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible in water
Formula Weight 214.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Naphthyl butyrate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'