missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl acetate, 99%, Thermo Scientific Chemicals

Catalog Number 10685092
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
830-81-9
C12H10O2
186.21
MFCD00003922
VGKONPUVOVVNSU-UHFFFAOYSA-N
1-naphthyl acetate, 1-acetoxynaphthalene, alpha-naphthyl acetate, 1-naphthol, acetate, a-naphthyl acetate, naphthyl acetate, naphthalene, 1-acetoxy, 1-naphthalenol, acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate
13247
naphthalen-1-yl acetate
CC(=O)OC1=CC=CC2=CC=CC=C21
This item is not returnable. View return policy
830-81-9
C12H10O2
186.21
MFCD00003922
VGKONPUVOVVNSU-UHFFFAOYSA-N
1-naphthyl acetate, 1-acetoxynaphthalene, alpha-naphthyl acetate, 1-naphthol, acetate, a-naphthyl acetate, naphthyl acetate, naphthalene, 1-acetoxy, 1-naphthalenol, acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate
13247
naphthalen-1-yl acetate
CC(=O)OC1=CC=CC2=CC=CC=C21

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 43.0°C to 46.0°C
Color Pink or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3CO2C10H7
Quantity 100 g
Beilstein 06, 608
Formula Weight 186.21
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-Naphthyl acetate, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'