missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoyl chloride, 99%, Thermo Scientific Chemicals

Catalog Number p-3758533
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
879-18-5
C11H7ClO
190.63
MFCD00004002
NSNPSJGHTQIXDO-UHFFFAOYSA-N
1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chloride
70146
naphthalene-1-carbonyl chloride
ClC(=O)C1=C2C=CC=CC2=CC=C1
This item is not returnable. View return policy
879-18-5
C11H7ClO
190.63
MFCD00004002
NSNPSJGHTQIXDO-UHFFFAOYSA-N
1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chloride
70146
naphthalene-1-carbonyl chloride
ClC(=O)C1=C2C=CC=CC2=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 16°C to 19°C
Color White
Density 1.2600g/mL
Boiling Point 190°C (35.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6515 to 1.6535
Quantity 10 g
Beilstein 09, 648
Specific Gravity 1.26
Solubility Information Solubility in water: decomposes
Formula Weight 190.63
Percent Purity 99%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material 1-Naphthoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-Naphthoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'