missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaleneboronic acid, 97%, ACROS Organics™

Brand:  Acros Organics 344690250

Code : 29

Additional Details : CAS Number : 13922-41-3 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10501744

  • £177.60 / 25g


Estimated Shipment
15-02-2018
Add to basket


Specifications

Chemical Identifiers

1-Naphthaleneboronic acid
96.0
97%
C10H9BO2
13922-41-3
100.0
97%
171.99

Specifications

1-Naphthaleneboronic acid
96.0
Authentic
Solubility in water: +/- 2.5% in water.
97%
C10H9BO2
196.0° to 200.0°C
25g
13922-41-3
100.0
Glass bottle
Powder
97%
171.99
Off-white to pink beige
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Naphthaleneboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS