missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-n-Octylpiperazine, 97%

Brand:  Alfa Aesar™ H55672.03

Code : NEW

Additional Details : CAS Number : 54256-45-0 Weight : 0.00100kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15451988

  • £36.50 / 1g


Estimated Shipment date
from Supplier 17-07-2018
Add to basket

Specifications

Chemical Identifiers

1-n-Octylpiperazine
198.35
UN1993
54256-45-0
MFCD00958407

Specifications

1-n-Octylpiperazine
240° to 241°C
1g
1.4670
97%
MFCD00958407
0.836g/mL
Colorless
54256-45-0
Liquid
198.35
UN1993
Videos
SDS