missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-n-Heptyl-4-(4-pyridyl)pyridinium bromide, 95%

Brand:  Alfa Aesar™ H55591.14

Code : NEW

Additional Details : CAS Number : 39127-10-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15421908

  • £503.50 / 25g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

1-n-Heptyl-4-(4-pyridyl)pyridinium bromide
39127-10-1
335.29
MFCD00134461

Specifications

1-n-Heptyl-4-(4-pyridyl)pyridinium bromide
125° to 128°C
39127-10-1
95%
MFCD00134461
White to yellow
25g
Powder
335.29
Videos
SDS