missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-n-Decyl-3-methylimidazolium tetrafluoroborate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
Description
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for pharmaceutical.
Notes
Moisture Sensitive, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is employed as a intermediate for pharmaceutical.
Notes
Moisture Sensitive, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Color | Yellow |
Refractive Index | 1.444 |
Quantity | 50 g |
Sensitivity | Moisture sensitive |
Formula Weight | 310.18 |
Percent Purity | 97% |
Physical Form | Liquid |
Chemical Name or Material | 1-n-Decyl-3-methylimidazolium tetrafluoroborate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'