missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
21.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.
Solubility
Miscible with acetone, acetonitrile, ethyl acetate, isopropyl alcohol and methylene chloride. Immiscible with hexane, toluene and water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.
Solubility
Miscible with acetone, acetonitrile, ethyl acetate, isopropyl alcohol and methylene chloride. Immiscible with hexane, toluene and water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Density | 1.21 |
Flash Point | None |
Refractive Index | 1.422 |
Quantity | 10 g |
Sensitivity | Hygroscopic |
Solubility Information | Miscible with acetone,acetonitrile,ethyl acetate,isopropyl alcohol and methylene chloride. Immiscible with hexane,toluene and water. |
IUPAC Name | 1-butyl-3-methylimidazol-3-ium;tetrafluoroborate |
Formula Weight | 226.03 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-n-Butyl-3-methylimidazolium tetrafluoroborate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'