missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
Description
1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.
Solubility
Insoluble in water.
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.
Solubility
Insoluble in water.
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Specifications
Color | Brown |
Refractive Index | 1.472 |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 348.42 |
Percent Purity | 96% |
Physical Form | Liquid |
Chemical Name or Material | 1-n-Butyl-3-methylimidazolium di-n-butyl phosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'