missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(Methylsulfonyl)piperazine, 97%

Brand:  Alfa Aesar™ H63870.22

Code : NEW

Additional Details : CAS Number : 55276-43-2 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15410819

  • £336.30 / 100g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Description

Description

Applications
1-(Methylsulfonyl)piperazine is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1-(Methylsulfonyl)piperazine
55276-43-2
164.23
MFCD02153225

Specifications

1-(Methylsulfonyl)piperazine
55276-43-2
97%
MFCD02153225
100g
Solid
164.23
Videos
SDS