missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals

Catalog Number 10478090
78.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
120-94-5
C5H11N
85.15
MFCD00003173
AVFZOVWCLRSYKC-UHFFFAOYSA-N
n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine
8454
1-methylpyrrolidine
CN1CCCC1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
120-94-5
C5H11N
85.15
MFCD00003173
AVFZOVWCLRSYKC-UHFFFAOYSA-N
n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine
8454
1-methylpyrrolidine
CN1CCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -90.0°C
Color Colorless
Density 0.8190g/mL
Boiling Point 80.0°C to 81.0°C
Flash Point −21°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4237 to 1.4257
Quantity 500 g
Beilstein 20,4
Specific Gravity 0.819
Solubility Information Solubility in water: fully miscible.
Viscosity 0.48 mPa.s (20°C)
Formula Weight 85.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Methylpyrrolidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'