Learn More
1-Methylpsilocin, Tocris Bioscience™
Potent and selective 5-HT2C agonist
Brand:  Tocris (Bio-Techne) 301750
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13462244
  • £505.72 / 50mg
Estimated Shipment
03-12-2018
Add to basket
Description
Description
Potent and selective 5-HT2C agonist (IC50 values are 12 and 633 nM for 5-HT2C and 5-HT2A receptors respectively). Displays high affinity for the 5-HT2B receptor (Ki = 38 nM) but acts as an inverse agonist. Active in vivo; inhibits scratching in a mouse model of OCD.
Specifications
Chemical Identifiers
1465-16-3
218.3
1-methylpsilocin, 3-2-dimethylamino ethyl-1-methylindol-4-ol, 3-2-dimethylamino-ethyl-1-methyl-1h-indol-4-ol, 3-2-dimethylamino ethyl-1-methyl-1h-indol-4-ol, 3-2-dimethylamino ethyl-1-methyl-1h-indol-4-ol, d0r8vt
3-[2-(dimethylamino)ethyl]-1-methylindol-4-ol
C13H18N2O
MZZRFEIDRWKTKJ-UHFFFAOYSA-N
44404883
CN1C=C(C2=C1C=CC=C2O)CCN(C)C
Specifications
1-Methylpsilocin
1465-16-3
1-methylpsilocin, 3-2-dimethylamino ethyl-1-methylindol-4-ol, 3-2-dimethylamino-ethyl-1-methyl-1h-indol-4-ol, 3-2-dimethylamino ethyl-1-methyl-1h-indol-4-ol, 3-2-dimethylamino ethyl-1-methyl-1h-indol-4-ol, d0r8vt
CN1C=C(C2=C1C=CC=C2O)CCN(C)C
218.3
218.29
50mg
C13H18N2O
MZZRFEIDRWKTKJ-UHFFFAOYSA-N
3-[2-(dimethylamino)ethyl]-1-methylindol-4-ol
44404883
>99%
Videos