missing translation for 'onlineSavingsMsg'
Learn More

1-Methylnaphthalene, 96%, Thermo Scientific Chemicals

Catalog Number p-164864
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
90-12-0
C11H10
142.2
MFCD00004034
QPUYECUOLPXSFR-UHFFFAOYSA-N
alpha-methylnaphthalene, naphthalene, 1-methyl, methylnaphthalene, naphthalene, methyl, polymethylnaphthalene, 1-methyl-naphthalene, alpha-methyl naphthalenes, methyl naphthalene, 1-methyl naphthalene, fema number 3193
7002
CHEBI:50717
1-methylnaphthalene
CC1=CC=CC2=CC=CC=C12
This item is not returnable. View return policy
90-12-0
C11H10
142.2
MFCD00004034
QPUYECUOLPXSFR-UHFFFAOYSA-N
alpha-methylnaphthalene, naphthalene, 1-methyl, methylnaphthalene, naphthalene, methyl, polymethylnaphthalene, 1-methyl-naphthalene, alpha-methyl naphthalenes, methyl naphthalene, 1-methyl naphthalene, fema number 3193
7002
CHEBI:50717
1-methylnaphthalene
CC1=CC=CC2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % >97
Melting Point -22.0°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 240.0°C to 243.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH3
Refractive Index 1.6130 to 1.6160
Quantity 100 g
Beilstein 05, 566
Specific Gravity 1.02
Solubility Information Solubility in water: 26mg/l (25°C). Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 142.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Methylnaphthalene, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methylnaphthalene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'