missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole, 98+%, Thermo Scientific Chemicals

Catalog Number p-3779124
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
603-76-9
C9H9N
131.18
MFCD00005800
BLRHMMGNCXNXJL-UHFFFAOYSA-N
1-methyl-1h-indole, n-methylindole, 1h-indole, 1-methyl, indole, 1-methyl, 1-methylindol, methylindole, unii-8h698roj5f, n-, chembl19912, n-methylindol
11781
1-methylindole
CN1C=CC2=CC=CC=C21
This item is not returnable. View return policy
603-76-9
C9H9N
131.18
MFCD00005800
BLRHMMGNCXNXJL-UHFFFAOYSA-N
1-methyl-1h-indole, n-methylindole, 1h-indole, 1-methyl, indole, 1-methyl, 1-methylindol, methylindole, unii-8h698roj5f, n-, chembl19912, n-methylindol
11781
1-methylindole
CN1C=CC2=CC=CC=C21

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Colorless to White
Density 1.0200g/mL
Boiling Point 236°C to 239°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.6060 to 1.608
Quantity 5 g
Specific Gravity 1.02
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,benzene
Formula Weight 131.18
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Methylindole
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methylindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'