missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline, 99+%, Thermo Scientific Chemicals

Catalog Number p-161713
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4721-98-6
C12H15NO2
205.26
MFCD00040276
VASUQTGZAPZKFK-UHFFFAOYSA-N
1-methyl-6,7-dimethoxy-3,4-dihydroisoquinoline, isoquinoline, 3,4-dihydro-6,7-dimethoxy-1-methyl, 6,7-dimethoxy-1-methyl-3,4-dihydro-isoquinoline, dehydrosalsolidine, maybridge1_001068, 3,4-dihydro-6,7-dimethoxy-1-methylisoquinoline, vasuqtgzapzkfk-uhfffaoysa, 1-methyl-3,4-dihydro-6,7-dimethoxyisoquinoline, 1-methyl-6?7-dimethoxy-3?4-dihydroisoquinoline, #
22652
6,7-dimethoxy-1-methyl-3,4-dihydroisoquinoline
CC1=NCCC2=CC(=C(C=C12)OC)OC
This item is not returnable. View return policy
4721-98-6
C12H15NO2
205.26
MFCD00040276
VASUQTGZAPZKFK-UHFFFAOYSA-N
1-methyl-6,7-dimethoxy-3,4-dihydroisoquinoline, isoquinoline, 3,4-dihydro-6,7-dimethoxy-1-methyl, 6,7-dimethoxy-1-methyl-3,4-dihydro-isoquinoline, dehydrosalsolidine, maybridge1_001068, 3,4-dihydro-6,7-dimethoxy-1-methylisoquinoline, vasuqtgzapzkfk-uhfffaoysa, 1-methyl-3,4-dihydro-6,7-dimethoxyisoquinoline, 1-methyl-6?7-dimethoxy-3?4-dihydroisoquinoline, #
22652
6,7-dimethoxy-1-methyl-3,4-dihydroisoquinoline
CC1=NCCC2=CC(=C(C=C12)OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 103°C to 106°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol,methanol,chloroform
Formula Weight 205.26
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-6, 7-dimethoxy-3, 4-dihydroisoquinoline, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'