missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol, 97%, Maybridge

Brand:  Maybridge BTB09194DA

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 119022-51-4 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10126423

  • £15.00 / 1g


Estimated Shipment date
from Supplier 03-08-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol
96.5
97%
C5H5F3N2O
MFCD00085094
119022-51-4
100.0
97%
166.1

Specifications

1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol
119022-51-4
100.0
97%
C5H5F3N2O
MFCD00085094
1g
96.5
Amber Glass Bottle
97%
166.1
Documentation

Technical Documents