missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole, 97%, Thermo Scientific™

Catalog Number p-3110113
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
191606-78-7
C17H31F3N2Sn
439.154
MFCD06797466
KZKSCFOBDCKLCL-UHFFFAOYSA-N
1-methyl-5-tributylstannyl-3-trifluoromethyl-1h-pyrazole, 1-methyl-3-trifluoromethyl-5-tributylstannylpyrazole, 1-methyl-5-tributylstannyl-3-trifluoromethyl pyrazole, tributyl-2-methyl-5-trifluoromethyl pyrazol-3-yl stannane, 1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl tributylstannane, 1h-pyrazole,1-methyl-5-tributylstannyl-3-trifluoromethyl, tributyl-2-methyl-5-trifluoromethyl-3-pyrazolyl stannane, 1-methyl-5 tributylstannyl 3-trifluoromethyl-1h-pyrazole, 1-methyl-5-tributylstannanyl-3-trifluoromethyl-1h-pyrazole
12056710
tributyl-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]stannane
CCCC[Sn](CCCC)(CCCC)C1=CC(=NN1C)C(F)(F)F
This item is not returnable. View return policy
191606-78-7
C17H31F3N2Sn
439.154
MFCD06797466
KZKSCFOBDCKLCL-UHFFFAOYSA-N
1-methyl-5-tributylstannyl-3-trifluoromethyl-1h-pyrazole, 1-methyl-3-trifluoromethyl-5-tributylstannylpyrazole, 1-methyl-5-tributylstannyl-3-trifluoromethyl pyrazole, tributyl-2-methyl-5-trifluoromethyl pyrazol-3-yl stannane, 1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl tributylstannane, 1h-pyrazole,1-methyl-5-tributylstannyl-3-trifluoromethyl, tributyl-2-methyl-5-trifluoromethyl-3-pyrazolyl stannane, 1-methyl-5 tributylstannyl 3-trifluoromethyl-1h-pyrazole, 1-methyl-5-tributylstannanyl-3-trifluoromethyl-1h-pyrazole
12056710
tributyl-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]stannane
CCCC[Sn](CCCC)(CCCC)C1=CC(=NN1C)C(F)(F)F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 439.15
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'