missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-5-tributylstannanyl-1H-imidazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 371.15 |
Percent Purity | 97% |
Chemical Name or Material | 1-Methyl-5-tributylstannanyl-1H-imidazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'