missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-5-phenyl-1H-pyrazol-3-yl)methylamine, 95%, Thermo Scientific™

Catalog Number p-3175582
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
869901-12-2
C11H13N3
187.246
MFCD08271937
QUPQCDFVZUNOJX-UHFFFAOYSA-N
1-methyl-5-phenyl-1h-pyrazol-3-yl methylamine, 1-1-methyl-5-phenyl-1h-pyrazol-3-yl methanamine, 1-methyl-5-phenylpyrazol-3-yl methanamine, 1h-pyrazole-3-methanamine,1-methyl-5-phenyl, 1-methyl-5-phenyl-1h-pyrazol-3-yl methanamine, 1-methyl-5-phenylpyrazol-3-yl methylamine, 1-1-methyl-5-phenylpyrazol-3-yl methanamine
18525794
(1-methyl-5-phenylpyrazol-3-yl)methanamine
CN1C(=CC(=N1)CN)C2=CC=CC=C2
This item is not returnable. View return policy
869901-12-2
C11H13N3
187.246
MFCD08271937
QUPQCDFVZUNOJX-UHFFFAOYSA-N
1-methyl-5-phenyl-1h-pyrazol-3-yl methylamine, 1-1-methyl-5-phenyl-1h-pyrazol-3-yl methanamine, 1-methyl-5-phenylpyrazol-3-yl methanamine, 1h-pyrazole-3-methanamine,1-methyl-5-phenyl, 1-methyl-5-phenyl-1h-pyrazol-3-yl methanamine, 1-methyl-5-phenylpyrazol-3-yl methylamine, 1-1-methyl-5-phenylpyrazol-3-yl methanamine
18525794
(1-methyl-5-phenylpyrazol-3-yl)methanamine
CN1C(=CC(=N1)CN)C2=CC=CC=C2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.24
Percent Purity 95%
Chemical Name or Material (1-Methyl-5-phenyl-1H-pyrazol-3-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'