missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3586613
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
921939-08-4
C12H9F3N2O3
286.21
MFCD06203565
JLFWKPKAZSVCKI-UHFFFAOYSA-N
1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenoxy-3-trifluoromethyl pyrazole-4-carboxylic acid
24229721
1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carboxylic acid
CN1N=C(C(C(O)=O)=C1OC1=CC=CC=C1)C(F)(F)F
This item is not returnable. View return policy
921939-08-4
C12H9F3N2O3
286.21
MFCD06203565
JLFWKPKAZSVCKI-UHFFFAOYSA-N
1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenoxy-3-trifluoromethyl pyrazole-4-carboxylic acid
24229721
1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carboxylic acid
CN1N=C(C(C(O)=O)=C1OC1=CC=CC=C1)C(F)(F)F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 286.21
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'