missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-piperidinemethanol, 97%, Thermo Scientific Chemicals

Catalog Number p-4229310
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20691-89-8
C7H15NO
129.2
KJZLJGZZDNGGCA-UHFFFAOYSA-N
1-methyl-4-piperidinemethanol, 1-methylpiperidin-4-yl methanol, n-methyl-4-piperidinemethanol, 4-piperidinemethanol, 1-methyl, 1-methyl-4-piperidinyl methanol, 4-hydroxymethyl-1-methylpiperidine, 1-methylpiperidine-4-methanol, n-methyl-4-hydroxymethylpiperidine, 1-methyl-piperidin-4-yl-methanol, 1-methyl-4-piperidyl methan-1-ol
271971
(1-methylpiperidin-4-yl)methanol
CN1CCC(CC1)CO
This item is not returnable. View return policy
20691-89-8
C7H15NO
129.2
KJZLJGZZDNGGCA-UHFFFAOYSA-N
1-methyl-4-piperidinemethanol, 1-methylpiperidin-4-yl methanol, n-methyl-4-piperidinemethanol, 4-piperidinemethanol, 1-methyl, 1-methyl-4-piperidinyl methanol, 4-hydroxymethyl-1-methylpiperidine, 1-methylpiperidine-4-methanol, n-methyl-4-hydroxymethylpiperidine, 1-methyl-piperidin-4-yl-methanol, 1-methyl-4-piperidyl methan-1-ol
271971
(1-methylpiperidin-4-yl)methanol
CN1CCC(CC1)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 29.0°C to 31.0°C
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 200.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Specific Gravity 0.98
Formula Weight 129.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Methyl-4-piperidinemethanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-4-piperidinemethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'