missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one, 96%, Thermo Scientific™

Catalog Number p-7081761
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1481-02-3
C5H5F3N2O
166.103
MFCD00051654
SYHYFYSGUQRNRS-UHFFFAOYSA-N
1-methyl-3-trifluoromethyl-2-pyrazolin-5-one, 1-methyl-3-trifluoromethyl-4,5-dihydro-1h-pyrazol-5-one, 2-methyl-5-trifluoromethyl-4h-pyrazol-3-one, 2-methyl-5-trifluoromethyl-2,4-dihydro-3h-pyrazol-3-one, 3h-pyrazol-3-one,2,4-dihydro-2-methyl-5-trifluoromethyl, acmc-20aoth, 2-methyl-5-trifluoromethyl-2,4-dihydro-pyrazol-3-one, 3-trifluoromethyl-1-methyl-1h-pyrazole-5 4h-one, 2-methyl-5-trifluoromethyl-2,4-dihydro-3h-pyrazol-3-one #
518566
2-methyl-5-(trifluoromethyl)-4H-pyrazol-3-one
CN1C(=O)CC(=N1)C(F)(F)F
This item is not returnable. View return policy
1481-02-3
C5H5F3N2O
166.103
MFCD00051654
SYHYFYSGUQRNRS-UHFFFAOYSA-N
1-methyl-3-trifluoromethyl-2-pyrazolin-5-one, 1-methyl-3-trifluoromethyl-4,5-dihydro-1h-pyrazol-5-one, 2-methyl-5-trifluoromethyl-4h-pyrazol-3-one, 2-methyl-5-trifluoromethyl-2,4-dihydro-3h-pyrazol-3-one, 3h-pyrazol-3-one,2,4-dihydro-2-methyl-5-trifluoromethyl, acmc-20aoth, 2-methyl-5-trifluoromethyl-2,4-dihydro-pyrazol-3-one, 3-trifluoromethyl-1-methyl-1h-pyrazole-5 4h-one, 2-methyl-5-trifluoromethyl-2,4-dihydro-3h-pyrazol-3-one #
518566
2-methyl-5-(trifluoromethyl)-4H-pyrazol-3-one
CN1C(=O)CC(=N1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 180°C
Quantity 1 g
Beilstein 5811070
Formula Weight 166.1
Percent Purity 96%
Chemical Name or Material 1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'